Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AZD 6482, Tocris Bioscience™

Catalog No. p-4817164 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective PI 3-Kβ inhibitor

Potent and selective inhibitor of PI 3-kinase β (PI 3-Kβ) (IC50 values are 0.69, 13.6, 47.8 and 136 nM for PI 3-Kβ, PI 3-Kδ, PI 3-Kγ and PI 3-Kα, respectively). Selectively inhibits in vivo growth of the PTEN-deficient tumor xenografts HCC70 and PC3. Also exhibits antithrombotic activity.

Chemical Identifiers

CAS 1173900-33-8
Molecular Formula C22H24N4O4
Molecular Weight (g/mol) 408.458
InChI Key IRTDIKMSKMREGO-OAHLLOKOSA-N
Synonym r-2-1-7-methyl-2-morpholino-4-oxo-4h-pyrido 1,2-a pyrimidin-9-yl ethylamino benzoic acid, 2-1r-1-7-methyl-2-4-morpholinyl-4-oxo-4h-pyrido 1,2-a pyrimidin-9-yl ethyl amino benzoic acid, unii-78g6mp5pz5, 2-1r-1-7-methyl-2-morpholin-4-yl-4-oxopyrido 1,2-a pyrimidin-9-yl ethyl amino benzoic acid, 4urk, pubchem19330, d0d0qa, azd 6482 who-dd
PubChem CID 44137675
IUPAC Name 2-[[(1R)-1-(7-methyl-2-morpholin-4-yl-4-oxopyrido[1,2-a]pyrimidin-9-yl)ethyl]amino]benzoic acid
SMILES CC1=CN2C(=O)C=C(N=C2C(=C1)C(C)NC3=CC=CC=C3C(=O)O)N4CCOCC4

Specifications

Quantity 10 mg
Formula Weight 408.45
Percent Purity >98%
Chemical Name or Material AZD 6482
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.