Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AZM 475271, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Src tyrosine kinase inhibitor. Reduces tumor size, vascularity and metastasis, and increases apoptosis in human pancreatic cells grown in nude mice. Also exhibits antiangiogenic activity in vitro and in vivo. Sensitizes tumor cells to the cytotoxic effects of gemcitabine (Cat. No. 3259).
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 442.94 |
Percent Purity | >99% |
Chemical Name or Material | AZM 475271 |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction