Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B-HT 933 dihydrochloride, Tocris Bioscience™

Catalog No. p-4783787 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective α2 agonist

Selective α2-adrenoceptor agonist. Exhibits greater than 300-fold selectivity for the α2-adrenoceptor over the α1-adrenoceptor.

Chemical Identifiers

CAS 36067-72-8
Molecular Formula C9H17Cl2N3O
Molecular Weight (g/mol) 254.16
MDL Number MFCD01529923
InChI Key HBLPYIOKPJVFQW-UHFFFAOYSA-N
Synonym b-ht 933 dihydrochloride, azepexole hydrochloride, azepexole dihydrochloride, 2-amino-6-ethyl-4,5,7,8-tetrahydro-6h-oxazolo 5,4-d azepine dihydrochloride, 5,6,7,8-tetrahydro-6-ethyl-4h-oxazolo 4,5-d azepin-2-amine dihydrochloride, 6-ethyl-5,6,7,8-tetrahydro-4h-oxazolo 4,5-d azepin-2-amine dihydrochloride, dsstox_cid_25756, dsstox_rid_81099, dsstox_gsid_45756, azepexole hcl
PubChem CID 169743
IUPAC Name dihydrogen 6-ethyl-4H,5H,6H,7H,8H-[1,3]oxazolo[4,5-d]azepin-2-amine dichloride
SMILES [H+].[H+].[Cl-].[Cl-].CCN1CCC2=C(CC1)N=C(N)O2

Specifications

Quantity 10 mg
Formula Weight 254.16
Percent Purity >99%
Chemical Name or Material B-HT 933 dihydrochloride
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.