Learn More
Alkali Scientific Baffled 125mL Erlenmeyer Flask with Vented Cap and Flat Bottom, PC, Sterile, 24/cs
SDP
Supplier:  Alkali Scientific EFB302
Encompass_Preferred
Baffled interior bottoms provide a swirl diruption/break which improves the aeration of the culture. This feature can be beneficial to use when the cultures' oxygen requirements are high or viscous.
Catalog No. 50-238-8964
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.