Learn More
Alkali Scientific Baffled 3000mL (3L) Erlenmeyer Flask with Vented Cap and Flat Bottom, PC, Sterile, 4/cs
Supplier: Alkali Scientific EFB308
Baffled interior bottoms provide a swirl diruption/break which improves the aeration of the culture. This feature can be beneficial to use when the cultures' oxygen requirements are high or viscous.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.