Learn More
Alkali Scientific Baffled 5000mL (5L) Erlenmeyer Flask with Vented Cap and Flat Bottom, PC, Sterile, 4/cs
SDP
Supplier:  Alkali Scientific EFB309
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Baffled interior bottoms provide a swirl diruption/break which improves the aeration of the culture. This feature can be beneficial to use when the cultures' oxygen requirements are high or viscous.
Catalog No. 50-238-8952