Learn More
Electron Microscopy Sciences AMD-SB Anti-Vibration Workstation AMD-SB Active Air Stainless Top, 120 x 75 cm, Stainless Steel Surface
Standard table has a white powder epoxy table top surface incorporating a AMT- 30x40 granite platform featuring four sorbothane isolators. It improves balance performance and is ideal for balances up to 4 to 5 digit.
The smooth seamless table shape ensures thorough and easy cleaning removing risks such as cross contamination. The stainless steel versions feature 240 satin silicon steel with a very low Ra of 0.4 making them ideal for cleanrooms and as well as being resistant to almost all forms of biological and chemical attack.The frame is produced using tubular ERW steel which combines high rigidity with a closed profile that allows easy and thorough cleaning. Tilting leveling feet ensure it sits perfectly in contact with the floor.
Size: 120 x 75 cm
Surface: Stainless Steel
Type: Active Air Self Leveling