Learn More
Medchemexpress LLC HY-100168 100mg Medchemexpress, BAPTA CAS:85233-19-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY100168100MG
Encompass_Preferred
Medchemexpress, HY-100168 100mg BAPTA CAS:85233-19-8 BAPTA is a calcium chelator. BAPTA suppresses intracellular reactive oxygen species ( ROS ) levels. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1736
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.