Learn More
Abbott Barbiturates (BAR) Urine Dip Test
Catalog No. 22911145
Click to view available options
:
A rapid, one step urine dip test for qualitative detection of barbiturates (BAR) in human urine
  • Detects barbiturates (BAR) at or above a cutoff of 300 ng/mL
  • Built in test control
  • Results in 5 min.
  • Results stable for up to 60 min.
Specifications
Detectable Analytes
BAR
Sample Type
Human Urine
Test Time
5 min.