Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Barium rod, 22mm (0.9 in.) dia. x 450mm (17.7 in.), 99+% (metals basis), Sr =0.8%, Thermo Scientific™
Click to view available options
Quantity:
1 Pc.
5 Pc.
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Packaged under argon
Specifications
Specifications
Color | Gray |
Quantity | 1 Pc. |
UN Number | UN1400 |
Percent Purity | ≥99% |
Concentration or Composition (by Analyte or Components) | Sr ≤0.8% |
Odor | Odorless |
Weight | ≈630g/rod |
Form | Rod |
Assay | (metals basis) |
Chemical Name or Material | Barium rod, 22mm (0.9 in.) dia. x 450mm (17.7 in.) |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS H Statement H228-H261 Flammable solid. In contact with water releases flammable gas. |
Precautionary Statement | GHS P Statement P210-P231+P232-P280-P370+P378i-P402+P404-P501a Keep away from heat/sparks/open flames/hot surfaces. No smoking. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. In case of fire: Use for extinction: Fire-extinguishing powder. Store in a dry place. Store in a closed container. Dispose of contents/container in accordance with local/regional/national/international regulations. |
DOTInformation | Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: BARIUM |
RTECSNumber | CQ8370000 |
TSCA | TSCA |
storageNote1 |
|
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction