Learn More
Medchemexpress LLC HY-107322 10mg Medchemexpress, Barnidipine (hydrochloride) CAS:104757-53-1 Purity:>98%
Medchemexpress, HY-107322 10mg Barnidipine (HCl) CAS:104757-53-1 Barnidipine hydrochloride (Mepirodipine hydrochloride) is an L-type calcium antagonist (CaA) with high affinity for [3H] initrendipine binding sites (Ki=0.21 nmol/l), has selective action against CaA receptors.Barnidipine hydrochloride (Mepirodipine hydrochloride) is an antihypertensive drug and acts by the reduction of peripheral vascular resistance secondary to its vasodilatory action. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.