Learn More
World Precision Instrument Barrier Milk Instrument Protector, Lubricant
SDP
Supplier:  World Precision Instrument 504215
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Barrier Milk Ready to Use Spray helps prevent corrosion of instruments when used prior to sterilization. Protects and extends the life of instruments during autoclaving. Simply spray the instrument directly before autoclaving with the unique water-based blend of lubricants and corrosion inhibitors. The fine mist works its way into hinges and box locks as well as covering all instruments with protection.
Catalog No. 50-253-9389
missing translation for 'QuantityLeft'