Learn More
Bioworld Glufosinate Ammonium, 500 mg
Supplier:  Bioworld 306320073
Encompass_Preferred
Specification:Synonym: Glufosinate ammonium, Phosphinothricin (PPT)Non-selective herbicide for plant transformation studies using nucleotide sequence of the phosphinothricin N-acetyltransferase gene from Streptomyces viridochromogenes, "Bar gene"Acts by inhibiting glutamine synthetase and thus blocks all nitrogen assimilation into the plantBasta is the registered trademark of Bayer CropScience
Catalog No. 50-255-087
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.