Learn More
Vector Laboratories Alpha-methylglucoside, 388 mg L-1280-5
Supplier: Vector Laboratories L12805
Alpha-methylglucoside, 388 mg. Alpha-methylglucoside sugar is provided for use as an inhibitor of lectin–conjugate binding or for eluting glycoproteins or other glycoconjugates from columns of agarose lectins. This sugar is supplied as a dry powder. When reconstituted in 5 ml of water, the resulting sugar solution is 400 mM.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.