Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAY 41-8543, >98%, Tocris Bioscience™

Catalog No. p-7121248 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Guanylyl cyclase (sGC) stimulator

Guanylyl cyclase (sGC) stimulator. Increases activity of sGC by up to 92-fold at concentrations up to 100 μM. Induces relaxation of aorta, saphenous arteries, coronary arteries and veins in vitro. Inhibits collagen-induced platelet aggregation. Promotes lipid uptake into brown adipose tissue and increases energy expenditure in mice. Induces weight loss in obese mice. Antihypertensive.

Chemical Identifiers

CAS 256498-66-5
Molecular Formula C21H21FN8O
Molecular Weight (g/mol) 420.452
InChI Key AQYFUZRYBJBAGZ-UHFFFAOYSA-N
Synonym unii-323t2c09sg, 2-1-2-fluorophenyl methyl-1h-pyrazolo 3,4-b pyridin-3-yl-5-4-morpholinyl-4,6-pyrimidinediamine, 4,6-pyrimidinediamine, 2-1-2-fluorophenyl methyl-1h-pyrazolo 3,4-b pyridin-3-yl-5-4-morpholinyl, 2-1-2-fluorophenyl methyl-1h-pyrazolo 3,4-b pyridin-3-yl-5-morpholin-4-yl pyrimidine-4,6-diamine, 2-1-2-fluorophenyl methyl pyrazolo 3,4-b pyridin-3-yl-5-morpholin-4-yl pyrimidine-4,6-diamine
PubChem CID 9953906
IUPAC Name 2-[1-[(2-fluorophenyl)methyl]pyrazolo[3,4-b]pyridin-3-yl]-5-morpholin-4-ylpyrimidine-4,6-diamine
SMILES C1COCCN1C2=C(N=C(N=C2N)C3=NN(C4=C3C=CC=N4)CC5=CC=CC=C5F)N

Specifications

Linear Formula C21H21FN8O
Quantity 10 mg
Solubility Information Soluble to 10mM in DMSO with gentle warming
Formula Weight 420.44
Percent Purity >98%
Chemical Name or Material 2-[1-[(2-Fluorophenyl)methyl]-1H-pyrazolo[3,4-b]pyridin-3-yl]-5-(4-morpholinyl)-4,6-pyrimidinediamin
Grade >98%
Recommended Storage Store at −20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.