Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BAY 58-2667 hydrochloride

Catalog No. 605210 Shop All R&D Systems Products
Click to view available options
:
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent soluble guanylyl cyclase (sGC) activator

BAY 58-2667 hydrochloride is a potent soluble guanylyl cyclase (sGC) activator (EC50 = 6.4 nM). Binds heme-free sGC. Exhibits antihypertensive effects in vivo.

Specifications

Chemical Name or Material

BAY 58-2667 hydrochloride

CAS

646995-35-9

Synonym

4-[[(4-Carboxybutyl)[2-[2-[[4-(2-phenylethyl)phenyl]methoxy]phenyl]ethyl]amino]methyl]benzoic acid hydrochloride

Quantity

10 mg

Solubility

Soluble to 100mM in DMSO

Molecular Formula

C36H39NO5.HCl

Formula Weight

602.16

Physical Form

Solid

Color

White

Product Type

Bioactive Small Molecule

Purity

>98%

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.