Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAY-u 3405, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent dual antagonist of TP/DP2 (CRTH2) prostanoid receptors (Ki values are 4.3, 4.5 and > 10000 nM for hDP2, hTP and hDP1 receptors respectively). Suppresses PGD2-induced migration of human eosinophils (IC50 = 170 nM).
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 416.47 |
Percent Purity | >99% |
Chemical Name or Material | BAY-u 3405 |
Safety and Handling
Recommended Storage | Desiccate at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction