Learn More
Brady™ BBP™31 Sign and Label Maker Accessory - Replacement Stylus Pens
Shop All Brady Corporation Products- Package includes three replacement Stylus Pens (printer sold separately)
- For use with BBP™31 Label Printer touch screen
- This stylus pen flows smoothly over the surface of the screen
- Navigation and selection are easy and accurate with a printer stylus
Specifications
Color
Blue
Description
Stylus
Autoclavable
Not Autoclavable
No. per Case
Pack of 5 Each
No. per Pack
Pack of 5 Each
Quantity
Pack of 5 Each
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.