Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ BBP™85 Series Printer Ribbon - R10000 Resin, Black/Blue/Green/Red

Catalog No. 19041117 Shop All Brady Corporation Products
Click to view available options
:

This industrial-grade ribbon is uniquely designed with memory cells that communicate information to the BBP™85 and PowerMark™ sign and label printers.

  • 8 in. panels of four different colors enable users to print four colors onto a label without having to swap ribbons
  • Tough, smear-proof ribbon produces long-lasting printed text that can withstand years in industrial areas and outdoor conditions
  • R10000 series ribbons are based on a resin formulation that offers a high performance image with strong solvent and scratch resistance
  • Ribbon cartridge eliminates calibration by automatically communicating information to the printer

Specifications

Product Type

Printer Ribbon

Color

Black/Blue/Green/Red

For Use With (Equipment)

BBP85 Industrial Sign and Label Printer, PowerMark Industrial Sign and Label Printer

Length (English)

200 ft.

Width (Metric)

92.71 mm

Autoclavable

Not Autoclavable

Description

Printer Ribbon

Format

Cartridge, Ribbon

Quantity

1 Roll

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.