Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BC 11 hydrobromide, Tocris Bioscience™

Catalog No. p-4816814 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective urokinase (uPA) inhibitor

Selective urokinase (uPA) inhibitor (IC50 = 8.2 μM). Exhibits no activity at 8 other related enzymes. Inhibits clot lysis with no effect on clot formation. Also decreases viability of MDA-MB231 breast cancer cells in vitro.

Chemical Identifiers

CAS 443776-49-6
Molecular Formula C8H12BBrN2O2S
Molecular Weight (g/mol) 290.97
InChI Key PAFZAMOVHIRQOD-UHFFFAOYSA-N
Synonym bc 11 hydrobromide, carbamimidothioic acid 4-boronophenyl methyl ester hydrobromide
PubChem CID 78243712
IUPAC Name [4-(carbamimidoylsulfanylmethyl)phenyl]boronic acid;hydrobromide
SMILES B(C1=CC=C(C=C1)CSC(=N)N)(O)O.Br

Specifications

Quantity 10 mg
Formula Weight 290.97
Percent Purity >98%
Chemical Name or Material BC 11 hydrobromide
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.