Learn More
Thomas Scientific Buchner Funnel with Medium Porosity Fixed Filter Plates, 10.25"
SDP
Supplier:  Thomas Scientific 1226D02
Encompass
Table-top Buchner funnels with medium porosity (45-90 microns) fixed filter plates are 6.4mm (1/4") thick high density polyethylene with a non-porous circumference to better fasten filter paper to the plate. The fixed plate models can be used for coarse filtration or with a cloth or paper filter. All plates are supported by a multiple ring grid to prevent displacement under vacuum. All Buchner funnels are rated for use below 52ºC (125ºF). Large volume, one piece design. Rugged and rigid polyethylene construction, ideal for heavy use.1226D02: 260mm (10.25") Inside Diameter, 178mm (7") Overall Height, 127mm (5")
Catalog No. NC2746434
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.