Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™/Difco™ Media Additives/Ingredients/Reagents: PALCAM Antimicrobic Supplement

Catalog No. DF063757 Shop All BD Diagnostic Systems Products
Click to view available options
:
PALCAM Antimicrobic Supplement, use with PALCAM Medium Base; BD Difco; 10mL
This item is not returnable. View return policy
This item is not returnable. View return policy

For PALCAM selective isolation of Listeria from foods

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage 2°C to 8°C
Additive PALCAM antimicrobic Supplement
Form Dehydrated
For Use With (Application) PALCAM medium base in preparing Palcam medium
Packaging Type Vial
Quantity 10 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.