Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Mycoflask™ Prepared Tubed Media: Bile Esculin Agar Slants
Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
10/Pk.
100/Pk.
For Use With (Application):
Isolation and cultivation of pathogenic and nonpathogenic fungi
Isolation and identification of microorganisms requiring prolonged incubation
Description
- For cultures requiring prolonged incubation
Specifications
Specifications
Certifications/Compliance | CE, COMPF |
Color | Beige |
Content And Storage | 2°C to 8°C |
Form | Solid |
Manufacturing Origin | United States |
Media Type | Bile Esculin Agar |
Formulation | Pancreatic digest of Gelatin 5g/L, Beef extract 3g/L, Oxgall 20g/L, Ferric Citrate 0.5g/L, Esculin 1g/L, Agar 14g/L |
For Use With (Application) | Isolation and cultivation of pathogenic and nonpathogenic fungi |
Packaging Type | Tube |
Quantity | 100/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction