Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Mycoflask™ Urea Agar Slants, Complete (Christensen) Prepared Media
Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
10/Pk.
100 Pack
Description
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
Specifications
Certifications/Compliance | CE, BAM, CCAM, ISO, USDA |
Color | Yellow |
Content And Storage | 2°C to 8°C |
Form | Liquid |
Manufacturing Origin | United States |
Media Type | Urea Agar |
Formulation | Pancreatic digest of Gelatin 1g/L, Dextrose 1g/L, Sodium Chloride 5g/L, Potassium Phosphate 2g/L, Urea 20g/L, Phenol red 0.012g/L, Agar 15g/L |
For Use With (Application) | For differentiation of organisms, especially Enterobacteriaceae, on basis of Urease production |
Packaging Type | Glass Tube with Cap |
Quantity | 100 Pack |
For in vitro diagnostic use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction