Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: Inhibitory Mold Agar (Deep Fill)

Catalog No. L98191 Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
100/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

Moderately selective medium used for the isolation of pathogenic fungi

  • Ready-to-use
  • Contains chloramphenicol
  • 100 x 10mm and 100 x 15mm-style plates
  • Deep-filled plates reduce effects of drying during prolonged incubation

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Beige
Content And Storage 2°C to 8°C
Form Solid
Manufacturing Origin United States
Media Type Inhibitory mold Agar
Formulation Pancreatic digest of Casein 3g/L, Peptic digest of Animal Tissue 2g/L, Yeast extract 5g/L, Dextrose 5g/L, Starch 2g/L, Dextrin 1g/L, Chloramphenicol 0.125g/L, Sodium Phosphate 2g/L, Magnesium Sulfate 0.8g/L, Ferrous Sulfate 0.04g/L, Sodium Chloride 0.04g/L, Manganese Sulfate 0.16g/L, Agar 15g/L
For Use With (Application) Pathogenic fungi
Packaging Type Plate
Quantity 100/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.