Learn More
BD BBL™ Vitamin K1-Hemin Solution
Shop All BD Diagnostic Systems Products- Used as a culture medium enrichment for anaerobic microorganisms.
- Vitamin K1-Hemin Solution is a ready-to-use solution. The solution cannot be heated and must be added aseptically in the proper amounts to media that have been autoclaved and cooled to 50-55°C.
- Vitamin K1-Hemin Solution is usually employed in prepared media at a final concentration of 5% for optimal results. Add Vitamin K1-Hemin Solution as required.
Specifications
Certifications/Compliance
ISO/DIS, 11014
Content And Storage
2°C to 8°C
Additive
Vitamin K1-hemin Solution
Form
Viscous
For Use With (Application)
Dehydrated culture media
Packaging Type
Tube
Quantity
10 mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.