Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Anaerobic Agar

Catalog No. DF0536174 Shop All BD Diagnostic Systems Products
Click to view available options
:
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit

General purpose medium for anaerobic bacteria

Specifications

Content And Storage

Refrigerate

Form

Dehydrated

Media Type

Anaerobic Agar

Product Type

Agar

Formulation

Pancreatic digest of Casein 20g/L, Sodium Chloride 5g/L, Dextrose 10g/L, Agar 20g/L, Sodium Thioglycollate 2g/L, Sodium Formaldehyde Sulfoxylate 1g/L, Methylene blue 2mg/L

For Use With (Application)

For anaerobic bacteria

Packaging Type

Bottle

Quantity

500 g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.