Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Modified Oxford Antimicrobic Supplement

Catalog No. DF0218605 Shop All BD Diagnostic Systems Products
Click to view available options
:
This item is not returnable. View return policy
This item is not returnable. View return policy

Applications:

Used with Oxford medium base

Specifications

Certifications/Compliance

ISO/DIS, 11014

Content And Storage

2°C to 8°C

Additive

Modified Oxford Antimicrobic Supplement

Form

Liquid

For Use With (Application)

Oxford medium base

Packaging Type

Bottle

Quantity

10 mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.