Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Pharmingen™ IHC Zinc Fixative

An alternative to formalin fixation of tissue for paraffin embedded sections

$86.40 - $225.00

Specifications

Color Clear
Content And Storage Room temperature
Product Type Fixative
For Use With (Application) Immunohistochemical staining
Form Liquid
View More Specs

Products 2
Catalog Number Mfr. No. Dilution Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Dilution Quantity Price Quantity & Availability  
BDB550523
View Documents
BD
550523
1 X 1000mL
Each for $86.40
missing translation for 'QuantityLeft'
Add to Cart
 
BDB552658
View Documents
BD
552658
10 X 500mL
Each for $225.00
missing translation for 'QuantityLeft'
Add to Cart
 
Description

Description

Ten percent neutral buffered formalin fixes tissue by cross-linking proteins, potentially causing a conformational change or “masking” of the antigenic epitope and leading to reduced or absent antibody reactivity. Different antigen retrieval techniques are used to unmask the epitope; however, many times it is not possible to retrieve the epitope and get optimal immunohistochemical staining. In these situations the use of IHC Zinc fixative, which is a milder fixative than formalin, is recommended.

  • Zinc-fixed paraffin embedded sections show very good immunostaining with many leukocytes markers and depict tissue morphology comparable to formalin-fixation
  • Use of antigen retrieval techniques on the zinc-fixed paraffin sections may further enhance the antibody reactivity in some cases

There are a few antigens that are not preserved by zinc-fixation. In these situations use of fresh frozen tissue for immunohistochemical staining is the only alternative.

Specifications

Specifications

Clear
Fixative
Liquid
Room temperature
Immunohistochemical staining
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.