Learn More
Electron Microscopy Sciences Beryllium Support Film, 25x25 mm
Supplier: Electron Microscopy Sciences 76030
A beryllium support film will reduce background interference for microanalysis to a minimum. It is particularly useful where analyses for Carbon or silicon are required and supports contain these elements cannot be used.
A deposition of 250 Angstroms thick Beryllium onto the 0.005" thick, 25x25 mm squares of a Cu substrate. The Be can be removed by dissolving the substrate in nitric acid (50:50). The Be film will then be removed from the acid, washed in distilled water and mounted on TEM grids.
Be support film is its very fine grain size which produces a very sharp ring pattern that can be used for in-situ calibration.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.