Learn More
ILEX LIFE SCIENCES LLC denovoMATRIX beadMATRIX - chemically-defined precoated microcarriers for MSCs culture, 30 g (3 x 10 g)
SDP
Supplier:  ILEX LIFE SCIENCES LLC M0101X3
Encompass_Preferred
Manufactured in Germany by denovoMATRIX, beadMATRIX are animal component-free, chemically-defined pre-coated microcarriers specially designed for bioreactor-scale expansion of mesenchymal stromal cells (MSCs) in serum-free and xeno-free media. COMPLIANT: Fulfilling pharma grade requirements. ACCELERATION: Application of beadMATRIX accelerates the proliferation of highly viable and therapeutically active MSCs up to 25% compared to the leading brand competitor. QUALITY: Harvested cells show MSC-specific characteristics as defined by the International Society of Cellular Therapy such as distinctive CD-marker expression and trilineage differentiation potential. In addition, they present immunomodulatory potential and angiogenic activity. COMPATIBLE: beadMATRIX has been successfully tested with different bioreactor setups and several cell culture media. beadMATRIX is ideally suited for use in shaker flasks, shaker bags, stirred tank or vertical wheel bioreactors.
Catalog No. 50-233-9538
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.