Learn More
Aqua Phoenix™ Becoming Banneker Kit Components
Shop All AquaPhoenix Scientific Products

Specifications
Product Type
Sidewalk chalk
Color
Assorted
Grade
K to 5
For Use With (Application)
Education Kit
Quantity
5 Pk.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.