Learn More
Fisherbrand™ Benchtop Extraction Kit
Benchtop Extraction Kits used for the capture and containment of particulate generated on the bench space.
$341.00 - $12030.00
Specifications
Color White
Material Aluminum
View More Specs
Products 5
Catalog Number Product Type Price Quantity & Availability  
Catalog Number Product Type Price Quantity & Availability  
15-050-086
View Documents
Standard Extraction Kit
Each for $10,890.00
Add to Cart
15-050-087
View Documents
Carbon Extraction Kit
Each for $12,030.00
Add to Cart
15-050-088
View Documents
Standard Filter
Each for $3,400.00
Add to Cart
15-050-089
View Documents
Carbon Filter
Each for $3,400.00
Add to Cart
15-050-090
View Documents
Filter Connector
Each for $341.00
Add to Cart
Description
Description
  • The Benchtop extraction kits include arms with hood or nozzle, fan with speed control and standard filter (particle, HEPA or gas), hose, connection details, and table bracket.
  • The speed of the fan is variably controlled by a speed controller.
  • Complete with hoses and extraction hoods.
  • The hood is made of clear plastic, which increases the visibility.
Specifications
Specifications
White
Aluminum