Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Benzamide 99.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
25 g
500 g
Specifications
Specifications
Melting Point | 128°C |
Color | White |
Boiling Point | 288°C |
Quantity | 25 g |
Formula Weight | 121.14 |
Percent Purity | ≥99.0% (N) |
Physical Form | Crystalline Powder |
Chemical Name or Material | Benzamide |
Safety and Handling
RTECSNumber | CU8700000 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction