Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzamidine Hydrochloride 96.0+%, TCI America™
SDP

Catalog No. p-7118301 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
100 g
500 g

Chemical Identifiers

CAS 1670-14-0
Molecular Formula C7H9ClN2
Molecular Weight (g/mol) 156.613
MDL Number MFCD00013025
InChI Key LZCZIHQBSCVGRD-UHFFFAOYSA-N
Synonym benzamidine hydrochloride, benzimidamide hydrochloride, benzenecarboximidamide hydrochloride, benzenecarboximidamide, monohydrochloride, benzamidinium chloride, benzamidine hcl, benzamidine, hydrochloride, amidinobenzene hydrochloride, benzamidine, monohydrochloride, amidinobenzene hydrochloride 1:1
PubChem CID 80289
IUPAC Name benzenecarboximidamide;hydrochloride
SMILES C1=CC=C(C=C1)C(=N)N.Cl

Specifications

Melting Point 171°C
Color White
pH 4.0 to 6.5 (50g/L 25°C)
Quantity 25 g
Formula Weight 156.61
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzamidine Hydrochloride
RTECSNumber CV6260000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.