Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzo[g]quinoxaline 98.0+%, TCI America™
SDP

Catalog No. p-7123605 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 260-50-4
Molecular Formula C12H8N2
Molecular Weight (g/mol) 180.21
MDL Number MFCD18449114
InChI Key XEMRLVBSKVCUDL-UHFFFAOYSA-N
PubChem CID 12886698
IUPAC Name benzo[g]quinoxaline
SMILES C1=CC=C2C=C3N=CC=NC3=CC2=C1

Specifications

Melting Point 127°C
Color Yellow
Quantity 200 mg
Formula Weight 180.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzo[g]quinoxaline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.