Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Benzofuran-3-boronic Acid (contains varying amounts of Anhydride), TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
Quantity | 1 g |
Synonym | benzofuran-3-ylboronic acid, benzofuran-3-boronic acid, 1-benzofuran-3-yl boronic acid, boronic acid, 3-benzofuranyl, acmc-1csxn, benzo b furan-3-boronic acid, benzofuran-3-yl boronic acid |
SMILES | OB(O)C1=COC2=CC=CC=C12 |
IUPAC Name | (1-benzofuran-3-yl)boronic acid |
Molecular Weight (g/mol) | 161.95 |
Formula Weight | 161.95 |
Physical Form | Crystalline Powder |
Chemical Name or Material | Benzofuran-3-boronic Acid (contains varying amounts of Anhydride) |
Safety and Handling
TSCA | No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction