Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Chloroacetate 97.0+%, TCI America™
SDP

Catalog No. p-7124882 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
250 g

Chemical Identifiers

CAS 140-18-1
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
MDL Number MFCD00157062
InChI Key SOGXBRHOWDEKQB-UHFFFAOYSA-N
Synonym benzyl chloroacetate, benzyl monochloracetate, acetic acid, chloro-, phenylmethyl ester, chloroacetic acid, benzyl ester, monochloroacetate, chloroacetic acid benzyl ester, benzyl-alpha-chloroacetate, phenylmethyl chloroacetate, acetic acid, chloro-, benzyl ester, phenylmethyl 2-chloroacetate
PubChem CID 8786
IUPAC Name benzyl 2-chloroacetate
SMILES C1=CC=C(C=C1)COC(=O)CCl

Specifications

Color Yellow
Boiling Point 132°C
Quantity 25 g
Formula Weight 184.62
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Chloroacetate
RTECSNumber AF8750000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.