Learn More
AdipoGen Benzylamine hydrochloride
SDP
Supplier:  AdipoGen CDXB0604G025
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Chemical. CAS 3287-99-8. Formula C7H9N . HCl. MW 143.6. Benzylamine is an aromatic amine used as a building block or intermediate for the production of organic fine chemicals and many pharmaceutical compounds. It can be prepared by the hydrogenation of benzonitrile and reductive amination of benzaldehyde using a metal catalyst. Benzylamine undergoes oxidative deamination reaction catalyzed by benzylamine oxidase and monoamine oxidase B and has been used as a substrate for the MAO-B enzyme. The hydrotropic salt BzCl facilitates molecular self-assembly of surfactant SDBS into unilamellar and multilamellar vesicles.
Catalog No. 50-256-1586