Learn More
TARGETMOL CHEMICALS INC BEPRIDIL HYDROCHLORIDE 50MG
Supplier:  TARGETMOL CHEMICALS INC T5391
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Bepridil hydrochloride (CERM 1978) is a calcium channel blocker and also inhibits Na+/Ca2+ exchange (NCX) sodium channels and cardiac sarcolemmal KATP channels. purity: 99%
Catalog No. 50-257-6688