Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bepridil hydrochloride, Tocris Bioscience™

Catalog No. 411750 Shop All R&D Systems Products
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Non-selective Ca2+ channel blocker; also NCX inhibitor

Calcium channel blocker. Also inhibits Na+/Ca2+ exchange (NCX), sodium channels and cardiac sarcolemmal KATP channels; opens mitochondrial KATP channels. Antiarrhythmic and antihypertensive; acts as a calcium sensitizer.

Chemical Identifiers

CAS 68099-86-5
Molecular Formula C24H35ClN2O
Molecular Weight (g/mol) 403.007
InChI Key JXBBWYGMTNAYNM-UHFFFAOYSA-N
Synonym bepridil hydrochloride, cordium, bepridil hcl, 1-isobutoxy-2-pyrrolidino-3-n-benzylanilino propane hydrochloride, 1-pyrrolidineethanamine, beta-2-methylpropoxy methyl-n-phenyl-n-phenylmethyl-, monohydrochloride, bepridil monohydrochloride, n-benzyl-n-3-isobutoxy-2-pyrrolidin-1-yl-propyl aniline hydrochloride, n-benzyl-beta-2-methylpropoxy methyl-n-phenylpyrrolidine-1-ethylamine monohydrochloride, prestwick_697, opera_id_594
PubChem CID 50088
ChEBI CHEBI:52712
IUPAC Name N-benzyl-N-[3-(2-methylpropoxy)-2-pyrrolidin-1-ylpropyl]aniline;hydrochloride
SMILES CC(C)COCC(CN(CC1=CC=CC=C1)C2=CC=CC=C2)N3CCCC3.Cl

Specifications

Quantity 50 mg
Formula Weight 403
Percent Purity >99%
Chemical Name or Material Bepridil hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.