Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Betulinic acid, Tocris Bioscience™

Catalog No. p-4816561 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Antitumor and anti-HIV agent; activates NF-κB. Also TGR5 agonist

Betulinic acid is a natural triterpenoid that displays anti-HIV and antitumor activity. Induces the production of reactive oxygen species (ROS) and activates NF-κB. Also a GPBA receptor partial agonist (EC50 = 1.04 μM, efficacy 83%). Betulinic acid suppresses areobic glycolysis by regulating the Cav-1/NF-κB/c-Myc pathway in and inhibits growth in breast cancer cells.

Chemical Identifiers

CAS 472-15-1
Molecular Formula C30H48O3
Molecular Weight (g/mol) 456.711
InChI Key QGJZLNKBHJESQX-FZFNOLFKSA-N
Synonym betulinic acid, mairin, betulic acid, ccris 6748, 3-hydroxylup-20 29-en-28-oic acid, 3beta-hydroxy-20 29-lupaene-28-oic acid, 3beta-hydroxy-lup-20 29-en-28-oic acid, lup-20 29-en-28-oic acid, 3-hydroxy-, 3beta, 1r,3as,5ar,5br,7ar,9s,11ar,11br,13ar,13br-9-hydroxy-1-isopropenyl-5a,5b,8,8,11a-pentamethyl-1,2,3,4,5,6,7,7a,9,10,11,11b,12,13,13a,13b-hexadecahydrocyclopenta a chrysene-3a-carboxylic acid, 1r,3as,5ar,5br,7ar,9s,11ar,11br,13ar,13br-9-hydroxy-5a,5b,8,8,11a-pentamethyl-1-prop-1-en-2-yl-1,2,3,4,5,6,7,7a,9,10,11,11b,12,13,13a,13b-hexadecahydrocyclopenta a chrysene-3a-carboxylic acid
PubChem CID 64971
ChEBI CHEBI:3087
IUPAC Name (1R,3aS,5aR,5bR,7aR,9S,11aR,11bR,13aR,13bR)-9-hydroxy-5a,5b,8,8,11a-pentamethyl-1-prop-1-en-2-yl-1,2,3,4,5,6,7,7a,9,10,11,11b,12,13,13a,13b-hexadecahydrocyclopenta[a]chrysene-3a-carboxylic acid
SMILES CC(=C)C1CCC2(C1C3CCC4C5(CCC(C(C5CCC4(C3(CC2)C)C)(C)C)O)C)C(=O)O

Specifications

Quantity 10 mg
Formula Weight 456.7
Percent Purity >98%
Chemical Name or Material Betulinic acid
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.