Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BGC 20-761, Tocris Bioscience™

Catalog No. 332610 Shop All R&D Systems Products
Click to view available options
:
10mg
This item is not returnable. View return policy
This item is not returnable. View return policy

High affinity 5-HT6 antagonist

Selective, high affinity 5-HT6 antagonist (Ki = 20 nM). Reverses the amnesic effects of scopolamine (Cat. No. 1414) and enhances memory consolidation in a rat model.

Specifications

CAS 17375-63-2
Molecular Formula C19H22N2O
Quantity 10 mg
Synonym unii-6g5f6esv5i, mpdt, 6g5f6esv5i, chembl7318, 5-methoxy-n,n-dimethyl-2-phenyl-1h-indole-3-ethanamine, 2-5-methoxy-2-phenyl-1h-indol-3-yl-n,n-dimethylethanamine, d0c5gf, gtpl71, 1h-indole-3-ethanamine, 5-methoxy-n,n-dimethyl-2-phenyl, indole, 3-2-dimethylamino ethyl-5-methoxy-2-phenyl
InChI Key VSGPGYWZVPDDSK-UHFFFAOYSA-N
SMILES CN(C)CCC1=C(NC2=C1C=C(C=C2)OC)C3=CC=CC=C3
IUPAC Name 2-(5-methoxy-2-phenyl-1H-indol-3-yl)-N,N-dimethylethanamine
Molecular Weight (g/mol) 294.398
PubChem CID 6918515
Formula Weight 294.39
Percent Purity >98%
Chemical Name or Material BGC 20-761
Show More Show Less
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.