Learn More
Medchemexpress LLC HY-18071 5mg Medchemexpress, BI-9627 CAS:1204329-34-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY180715MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Medchemexpress, HY-18071 5mg BI-9627 CAS:1204329-34-9 BI-9627 is a sodium-hydrogen exchanger isoform 1 (NHE1) inhibitor with an EC 50 of 31 nM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2807
missing translation for 'QuantityLeft'