Learn More
Gold Biotechnology Inc Bialaphos, Sodium Salt 100 mg
SDP
Supplier:  Gold Biotechnology Inc B0178100
Encompass
Bialaphos is a tripeptide antibiotic that is used in transformation experiments of many species of plants that make use of the bar gene for selection. In corn, it is more effective than glufosinate. In wheat, bialaphos was the most reliable selection criteria used to identify bar transformed plants.
Catalog No. NC2942814
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.