Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bianthrone 95.0+%, TCI America™
SDP

Catalog No. p-7118694 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
100 mg
1 g
5 g

Chemical Identifiers

CAS 434-85-5
Molecular Formula C28H16O2
Molecular Weight (g/mol) 384.434
MDL Number MFCD00001238
InChI Key MGRRGKWPEVFJSH-UHFFFAOYSA-N
Synonym bianthrone, 10h,10'h-9,9'-bianthracenylidene-10,10'-dione, dehydrodianthrone, 9 10h-anthracenone, 10-10-oxo-9 10h-anthracenylidene, .delta.10,10'-bianthrone, 10-10-oxo-9-anthrylidene anthracen-9-one, 10-10-oxoanthracen-9-ylidene anthracen-9-one, bisanthrone, 10-10-oxoanthracen-9 10h-ylidene anthracen-9 10h-one, 9,9'-bianthracenylidene-10,10'-dione
PubChem CID 67948
IUPAC Name 10-(10-oxoanthracen-9-ylidene)anthracen-9-one
SMILES C1=CC=C2C(=C1)C(=C3C4=CC=CC=C4C(=O)C5=CC=CC=C53)C6=CC=CC=C6C2=O

Specifications

Melting Point 300°C
Color Green-Yellow
Quantity 100 mg
Formula Weight 384.43
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Bianthrone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.