Learn More
Apexbio Technology LLC (+)-Bicuculline,100mgCAS# 485-49-4
SDP
Supplier:  Apexbio Technology LLC N1592100
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
GABAA receptor antagonist; blocks Ca2+-activated potassium (SK) channels. Water soluble derivative of (+)-bicuculline. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0002