Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Bicuculline methiodide, Tocris Bioscience™

Catalog No. p-4783653 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

GABAA antagonist; more water soluble version of (+)-bicuculline (Cat. No. 0130)

Methiodide form of classical GABAA receptor antagonist (+)-bicuculline . More water-soluble and stable. Non-GABA receptor-mediated actions reported, including actions on calcium-dependent potassium channels.

Methobromide Salt and Methochloride Salt also available.

Chemical Identifiers

CAS 40709-69-1
Molecular Formula C21H20INO6
Molecular Weight (g/mol) 509.296
InChI Key HKJKCPKPSSVUHY-GRTNUQQKSA-M
Synonym --bicuculline methiodide, bicuculline methiodide, 1 s ,9 r---bicuculline methiodide, 5s-5-6r-6,8-dihydro-8-oxofuro 3,4-e-1,3-benzodioxol-6-yl-5,6,7,8-tetrahydro-6,6-dimethyl-1,3-dioxolo 4,5-g isoquinolinium iodide, 1,3-dioxolo 4,5-g isoquinolinium, 5-6r-6,8-dihydro-8-oxofuro 3,4-e-1,3-benzodioxol-6-yl-5,6,7,8-tetrahydro-6,6-dimethyl-, iodide, 5s, 1,3-dioxolo 4,5-g isoquinolinium, 5-6,8-dihydro-8-oxofuro 3,4-e-1,3-benzodioxol-6-yl-5,6,7,8-tetrahydro-6,6-dimethyl-, iodide, r-r*,s*, opera_id_515, --bicucullinemethiodide, 1 s ,9 r---bicuculline methiodide hpce, 5r-5-6s-6,8-dihydro-8-oxofuro 3,4-e-1,3-benzodioxol-6-yl-5,6,7,8-tetrahydro-6,6-dimethyl-1,3-dioxolo 4,5-g isoquinolinium iodide
PubChem CID 104871
IUPAC Name (6R)-6-[(5S)-6,6-dimethyl-7,8-dihydro-5H-[1,3]dioxolo[4,5-g]isoquinolin-6-ium-5-yl]-6H-furo[3,4-g][1,3]benzodioxol-8-one;iodide
SMILES C[N+]1(CCC2=CC3=C(C=C2C1C4C5=C(C6=C(C=C5)OCO6)C(=O)O4)OCO3)C.[I-]

Specifications

Quantity 10 mg
Formula Weight 509.3
Percent Purity >99%
Chemical Name or Material (-)-Bicuculline methiodide
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.