Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bifemelane hydrochloride, Tocris Bioscience™

Catalog No. p-4783283 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

MAO-A and MAO-B inhibitor

Bifemelane hydrochloride is an antidepressant MAO inhibitor. Reverses catalepsy induced by Tetrabenazine (Cat. No. 2175) in mice and increases locomotor activity in MPTP-treated marmosets.

Chemical Identifiers

CAS 62232-46-6
Molecular Formula C18H24ClNO
Molecular Weight (g/mol) 305.846
InChI Key MEAHDXWXNNDSAK-UHFFFAOYSA-N
Synonym bifemelane hydrochloride, celeport, alnert, unii-2n52s01ufy, 4-2-benzylphenoxy-n-methylbutan-1-amine hydrochloride, n-methyl-4-2-phenylmethyl phenoxy-1-butanamine hydrochloride, regid_for_cid_6917789, 4-2-benzylphenoxy-n-methylbutylamine hydrochloride
PubChem CID 6917789
IUPAC Name 4-(2-benzylphenoxy)-N-methylbutan-1-amine;hydrochloride
SMILES CNCCCCOC1=CC=CC=C1CC2=CC=CC=C2.Cl

Specifications

Quantity 10 mg
Formula Weight 305.84
Percent Purity >99%
Chemical Name or Material Bifemelane hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.