Learn More
ATCC BIG-6 ESCHERICHIA COLI STRAINS
Supplier:  ATCC MP9
Encompass
MP-9™, Big-Six Escherichia coli Strains PanelA panel of six of the Big-Six non-O157 STEC serogroups strains that provide a robust combination of Shiga toxin gene expression with varied sources of isolation for your research.
Catalog No. NC2204614
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.